Learn More
Thermo Scientific™ Forma™ Inner Door Kits
Shop All Thermo Scientific Products
Click to view available options
For Use With (Equipment):
Forma 7000 and 900 Series, 17.3 cu. ft. Uprights
Forma 7000 and 900 Series, 23.0 cu. ft. Uprights
Forma 7000 and 900 Series, 28.0 cu. ft. Uprights
Unit Size:
Each
Description
Reduce cold air loss and improve temperature recovery after door closings with Thermo Scientific™ Forma™ Inner Door Kits, a factory-installed door alternative option for your ultra-low upright freezer.
Four interior doors are already included on our -86°C uprights. The five inner door option for 8600 and 900 Series includes five powder-coated inner doors and one additional shelf.
Ordering Information:
Factory installed. Must be ordered with the refrigerator or freezer.
Spécification
Spécification
| Type | Ultra-Low Freezer Inner-Door |
| Capacity (English) | 17.3 cu. ft. |
| Capacity (Metric) | 489.8 L |
| Temperature Range | -86°C |
| No. of Doors | 5 |
| Exterior Finish | Powder-coated Steel |
| Shelves | 1 |
| Style | Upright |
| For Use With (Equipment) | Forma 7000 and 900 Series, 17.3 cu. ft. Uprights |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu