Learn More
Thermo Scientific™ Forma™ Freezer Replacement Shelves
Product Code. p-4530834 Shop All Thermo Scientific Products
Change view
Click to view available options
For Use With (Equipment):
13.0 cu. ft. Freezers
17.3 cu. ft. Freezers
23.0 cu. ft. Freezers
28 cu. ft. Freezers
Type:
Shelf Kit
Unit Size:
Each
Product Code. For Use With (Equipment) Type unitSize
11786469 13.0 cu. ft. Freezers Shelf Kit Each
11756479 17.3 cu. ft. Freezers Shelf Kit Each
11766479 23.0 cu. ft. Freezers Shelf Kit Each
11786649 28 cu. ft. Freezers Shelf Kit Each
4 options
Product Code. 11786469
Brand: Thermo Scientific™ 224134
Only null left
Add to basket
Add to basket
Use Thermo Scientific™ Forma™ Freezer Replacement Shelves with Forma 8600 and 900 Series Upright Freezers.
  • Stainless steel
  • Kits include mounting hardware
TRUSTED_SUSTAINABILITY
Specifications
Type Shelf Kit
Capacity (English) 13 cu. ft.
Capacity (Metric) 368.1 L
Style Upright
Color Silver
For Use With (Equipment) 13.0 cu. ft. Freezers