Description
- Stainless steel (1.4301)
- The pointed, fluted ends can be used to work reliably with sensitive materials, tissue or samples
- Flutings on the handle ensure a slip-free grip of the stainless steel tweezers
- Suitable for use in chemistry, the pharmaceutical industry, cosmetics, and food/beverage
Specifications
Specifications
| Product Type | Forceps |
| Length (English) | 5.12 in. |
| Length (Metric) | 130 mm |
| Material | Stainless Steel (1.4301) |
| For Use With (Application) | Food |
| No. per Pack | 1 |
| Style | Standard |
| Tip Style | Sharp, Bent |