Learn More
Honeywell™ FORCE (i)XTREM S3 Shoes
Product Code. 15876575
Change view
Click to view available options
Size:
37
38
39
40
41
42
43
44
45
46
47
48
Unit Size:
1 pair
Product Code. Size unitSize
15876575 45 1 pair
15896565 37 1 pair
15806575 38 1 pair
15816575 39 1 pair
15826575 40 1 pair
15836575 41 1 pair
15846575 42 1 pair
15856575 43 1 pair
15866575 44 1 pair
15886575 46 1 pair
15896575 47 1 pair
15806585 48 1 pair
12 options
Product Code. 15876575
Brand: Honeywell™ 624620345/7
Only null left
Add to basket
Add to basket
Honeywell™ FORCE (i)XTREM S3 Shoes are constructed of water-resistant full grain leather.
  • Water resistant grain full leather
  • Ergonomic collar and padded tongue for an optimized comfort
  • Gusset to avoid water and dust penetration
  • Breathable 3D lining
  • Derby style for a better fitting in all sizes
  • Toe cap metal free
  • Metal free antiperforation insert
  • Injected PU / Nitrile sole
  • (i)NTELLIGENT wear control
  • EN ISO 20345:2011 S3 HRO HI CI SRC
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance EN ISO 20345:2011 S3 HRO HI CI SRC
Color Black
Material Leather
Size 45
Sole Material Polyurethane and nitrile
Toe Style Metal-free