Learn More
Eppendorf™ Drawer Racking for U410 Model Upright ULT Freezers
Description
- Product Type: Drawer Racking for Upright ULT Freezers
- Material: Aluminum
- Dimensions (D x W x H): 563 mm x 140 mm x 231 mm
- Box Footprint: 133 mm x 133 mm
- Box Height: 53 mm
- Description: Contains (max.) 16 freezer boxes, 4 boxes per drawer, 4 drawers
- For Use With (Equipment): CryoCube™ F570 Series, CryoCube™ F440 Series, U410 Series, and CryoCube™ F740 Series (5-compartment) Upright Freezers
Specifications
Specifications
| Autoclavable | Autoclavable |
| Holds | Cryoboxes (133 x 133 x 53 mm) |
| Material | Aluminum |
| Positions | 16 |
| Stackable | No |
| Sterility | Non-sterile |
| For Use With (Application) | Cold Storage |
| For Use With (Equipment) | CryoCube™ F570 Series, CryoCube™ F440 Series, U410 Series, and CryoCube™ F740 Series (5-compartment) Upright Freezers |
| No. of Racks | 1 |
| Quantity | 1 Each |
| Show More |