missing translation for 'onlineSavingsMsg'
Learn More

Folinic acid, calcium salt, 99.4%, MP Biomedicals™

Product Code. 11454742 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
25 mg
0.1 g
0.5 g
1 g
Unit Size:
100mg
1g
25mg
500mg
Product Code. Quantity unitSize
11454742 0.1 g 100mg
11444742 25 mg 25mg
11464742 0.5 g 500mg
11474742 1 g 1g
4 options
This item is not returnable. View return policy

Product Code. 11454742

Brand: MP Biomedicals 0219150480

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 1492-18-8
Molecular Formula C20H21CaN7O7
Molecular Weight (g/mol) 511.51
MDL Number MFCD00006704
InChI Key KVUAALJSMIVURS-AEQORCIRNA-L
Synonym calcium folinate
PubChem CID 131675590
SMILES [Ca++].NC1=NC(=O)C2=C(NCC(CNC3=CC=C(C=C3)C(=O)N[C@@H](CCC([O-])=O)C([O-])=O)N2C=O)N1

Specifications

Color Yellow
Assay Percent Range 0.994
Quantity 0.1 g
IUPAC Name calcium (2S)-2-[(4-{[(2-amino-5-formyl-4-oxo-1,4,5,6,7,8-hexahydropteridin-6-yl)methyl]amino}phenyl)formamido]pentanedioate
Formula Weight 511.5
Physical Form Powder
Chemical Name or Material Folinic acid, calcium salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.