Learn More
ZVG Zellstoff Vertrieb™ Folded Towels
Product Code. p-8000811
Change view
Click to view available options
Dimensions (L x W):
25 x 21 cm
Ply:
2-ply
Unit Size:
Pack of 3200
Product Code. Dimensions (L x W) Ply unitSize
10771283 25 x 21 cm 2-ply Pack of 3200
1 options
Product Code. 10771283
Brand: ZVG Zellstoff Vertrieb™ 1542204
Only null left
Add to basket
Add to basket
Use ZVG Zellstoff Vertrieb™ Folded Towels in either 2-ply or 3-ply folded paper forms. They are available in various sizes, quantities and colors as well.
  • ZetForm™l paper towel system
  • Various colors
  • Various thicknesses
Specifications
Product Type Folded Towel
Length (Metric) 25 cm
Width (Metric) 21 cm
Material Paper
Dimensions (L x W) 25 x 21 cm
Ply 2-ply