Learn More
Keystone Cleanroom Products Cleanroom Face Masks, Gamma Irradiated with Tyvek™ Side Extension
BIOPHARMAGREENER_CHOICE
Product Code. 17755299 Shop All Keystone Cleanroom Products Products
Change view
Click to view available options
Fastening Type:
Headband
Ties
Color:
Blue
Green
Unit Size:
Pack of 500
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Fastening Type Color unitSize
17755299 Headband Blue Pack of 500
17765299 Headband Green Pack of 500
17775299 Ties Blue Pack of 500
17785299 Ties Green Pack of 500
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 17755299 Supplier Keystone Cleanroom Products Supplier No. S1461XLDEL2
  • ONLINE EXCLUSIVE PRICE
    37830.00 NOK
    29221.61 NOK / Pack of 500
    Save 8608.39 NOK
    22% Off
View all promotions
Estimated Shipment: 27-11-2026
Add to basket
Add to basket
Sterile cleanroom facemasks feature an tyvek side extension for additional protection. Available with ties or double elastic headband fastening.
  • Sterile cleanroom face masks
  • Certificates of Irradiation and Conformance with each shipment
  • Tyvek™ side extensions allow mask expansion to increase coverage and prevent unwanted facial skin exposure
Specifications
Product Type Cleanroom Face Mask
Color Blue
Fastening Type Headband
Material SMS
Size X-Large
Sterility Sterile
Dimensions (L x W) 12.25 x 4 in.
Style Pleated
Cleanroom Class Class 100 (ISO 5)
Disposable Single-use
Includes Side Extension
Latex Free Yes
Length (English) 12.25 in.
Length (Metric) 31 cm
Model S-1461
Width (English) 4 in.
Width (Metric) 10 cm