Learn More
Foamtec Stainless Steel Corrosion Remediation Kits
Product Code. 15123514
Encompass
Change view
Click to view available options
Product Type:
Large Surface Starter Kit
Natural Sahara Sponge
Small Surface Starter Kit
Unit Size:
Each
Product Code. Product Type unitSize
15123514 Natural Sahara Sponge Each
15103514 Large Surface Starter Kit Each
15183504 Small Surface Starter Kit Each
3 options
Product Code. 15123514
Brand: Foamtec HS48541
This item requires a price update prior to being ordered.
Request a Price for this item here.
Enable technicians to quickly remediate corrosion using a polishing and cleaning process that is fully compatible with cleanroom operations
Kits include polishing and cleaning products along with instructional videos and documentation that result in mirror smooth 304 and 316 SS surfaces that resist subsequent corrosion.
Kit Contents
19-304-555: Stainless Steel Corrosion Remediation Starter Kit for Large Surfaces
2 Handles, 10 Diamond ScrubDISKS (5 girts), 1 Silicon-Carbide ScrubDISK, 1PilloWIPE, 4 Diamond ScrubPADS (4 girts), 1 Silicon-Carbide ScrubPAD, ScrubWRIGHT Pen with 2 Belts, 25 MiraWIPE Wipers, 2 UltraSOLV Sponge, and Flash Drive with instructional Videos
19-304-556: Stainless Steel Corrosion Remediation Starter Kit
1 PilloWIPE, 4 Diamond ScrubPADS (4 girts), 1 Silicon-Carbide ScrubPAD, ScrubWRIGHT Pen with 2 Belts, 10 MiraWIPE Wipers, 1 UltraSOLV Sponge, and Flash Drive with instructional Videos
Specifications
Product Type Natural Sahara Sponge
For Use With (Application) Cleaning corrosion on stainless steel surfaces
For Use With (Equipment) Semi-Conductor Fabs, Pharmaceutical Cleanrooms, Medical Device Facilities