Learn More
Foamtec MiraWIPE™ Wipers
Description
MiraWIPE™ wipers feature a non-symmetrical micro fiber structure allows them to scrub microbial biofilms and sporicidal residues.
- Made of continuous filament micro denier, super soft polyester/nylon textile
- High absorbency and contamination pickup
- Abrasion resistant, no-run, no-stretch fabric to minimize In-Use Particles from the face of the wiper
- Available in configurations to meet Class 1-1000 applications
Specifications
Specifications
| Product Type | Cleanroom Wipe |
| Length (English) | 9 in. |
| Length (Metric) | 22.86 cm |
| Width (English) | 9 in. |
| Width (Metric) | 22.86 cm |
| Material | Polyester |
| Certifications/Compliance | ISO Class 4 |
| Edges | Extra wide sealed |
| Dimensions (L x W) | 9 x 9 in. (22.86 x 22.86 cm) |
| For Use With (Application) | Cleanroom |
| Show More |