Learn More
OHAUS™ Foam Test Tube Rack
Shop All Ohaus Products
Click to view available options
Holds:
10 mm Tubes
12 mm Tubes
13 mm Tubes
15 x 50 mL Tubes
16 mm Tubes
25 mm Tubes
Color:
Blue
Gray
Green
Red
White
Yellow
Unit Size:
Each
Description
- Constructed of foam
- For use with tubes of various sizes
Specifications
Specifications
| Holds | 15 x 50 mL Tubes |
| Product Type | Test Tube Rack |
| Material | Foam |
| Dimensions (L x W) | 140 x 241 mm |
| Color | Red |
| Description | 50 mL Tube Rack (Red) |
| No. of Positions | 15 |