missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ FMR1NB Recombinant Protein

Human FMR1NB full-length ORF recombinant protein with GST-tag at N-terminal

Brand:  Abnova™ H00158521-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16188853

  • 6245.00 NOK / 25µg
Estimated Shipment: 02-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • Theoretical MW (kDa): 53.57
  • Preparation method: In vitro wheat germ expression system
  • Purification: Glutathione Sepharose 4 fast flow
  • Storage buffer: 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer

Sequence: MSSHRRKAKGRNRRSHRAMRVAHLELATYELAATESNPESSHPGYEAAMADRPQPGWRESLKMRVSKPFGMLMLSIWILLFVCYYLSYYLCSGSSYFVLANGHILPNSENAHGQSLEEDSALEALLNFFFPTTCNLRENQVAKPCNELQDLSESECLRHKCCFSSSGTTSFKCFAPFRDVPKQMMQMFGLGAISLILVCLPIYCRSLFWRSEPADDLQRQDNRVVTGLKKQRRKRKRKSEMLQKAARGREEHGDE

Best use within three months from the date of receipt of this protein.

ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array

Specifications

Specifications

AAH34320
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
53.57
8
Glutathione Sepharose 4 Fast Flow
25 ug
MSSHRRKAKGRNRRSHRAMRVAHLELATYELAATESNPESSHPGYEAAMADRPQPGWRESLKMRVSKPFGMLMLSIWILLFVCYYLSYYLCSGSSYFVLANGHILPNSENAHGQSLEEDSALEALLNFFFPTTCNLRENQVAKPCNELQDLSESECLRHKCCFSSSGTTSFKCFAPFRDVPKQMMQMFGLGAISLILVCLPIYCRSLFWRSEPADDLQRQDNRVVTGLKKQRRKRKRKSEMLQKAARGREEHGDE
FLJ25736/NY-SAR-35
FMR1NB
Wheat Germ (in vitro)
GST
Antibody Production, Protein Array, ELISA, Western Blot
158521
FMR1NB (Human) Recombinant Protein (P01)
In vitro wheat germ expression system
12.5% SDS-PAGE Stained with Coomassie Blue.
Wheat Germ (in vitro)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FMR1NB
Human
Recombinant
Solution
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers