missing translation for 'onlineSavingsMsg'
Learn More

FMO4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 15394-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16894544

  • 1281.92 NOK / 20µL
Estimated Shipment: 15-12-2022
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This antibody, generated by N-330 amino-acids of FMO4, can recognize the 2 variants and also cross-react with other FMO proteins.

FMO4 (Flavin-containing monooxygenase 4) also known as FMO2, FORMERLY or FMO, ADULT LIVER FORM, is an enzyme that in humans is encoded by the FMO4 gene. By PCR analysis, Dolphin et al. (1992) mapped the FMO4 gene to chromosome 1. Southern blot hybridization with single exon probes demonstrated that human FMO4 and FMO1 are, in each case, the product of a single gene. Lawton et al. (1994) proposed a reclassification of mammalian FMOs. Under this system, FMO2 described by Dolphin et al. (1992) is now designated FMO4.
Specifications

Specifications

FMO4
Polyclonal
Unconjugated
FMO4
Dimethylaniline oxidase 4, FMO 4, FMO2, FMO4
Rabbit
Antigen Affinity Chromatography
RUO
226564, 2329, 246247
-20°C
Liquid
Western Blot
0.2 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P31512, Q8K4B7, Q8VHG0
Fmo4
FMO4 Fusion Protein Ag7590
20 μL
Primary
Rat, Mouse, Human
Antibody
IgG
Videos
SDS
Documents
Certificates
Special Offers

Special Offers