missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ FMNL2 Recombinant Protein

Product Code. p-3719022
Change view
Click to view available options
Quantity:
10 μg
25 μg
Unit Size:
10µg
25µg
Product Code. Quantity unitSize
16117113 10 μg 10µg
16127113 25 μg 25µg
2 options
This item is not returnable. View return policy

Product Code. 16117113

Brand: Abnova™ H00114793P01.10ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array

Specifications

Accession Number AAH36492
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
Gene ID (Entrez) 114793
Molecular Weight (g/mol) 45.32
Name FMNL2 (Human) Recombinant Protein (P01)
pH Range 8
Preparation Method In vitro wheat germ expression system
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 μg
Source Wheat Germ (in vitro)
Immunogen MDLTKREYTMHDHNTLLKEFILNNEGKLKKLQDDAKIAQDAFDDVVKYFGENPKTTPPSVFFPVFVRFVKAYKQAEEENELRKKQEQALMEKLLEQEALMEQQDPKSPSHKSKRQQQELIAELRRRQVKDNRHVYEGKDGAIEDIITALKKNNITKFPNVHSRVRISSSTPVVEDTQS
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Gene Alias FHOD2/FLJ37546
Common Name FMNL2
Gene Symbol FMNL2
Cross Reactivity Human
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Form Solution
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.