missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FMN1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25982-1-AP-150UL
This item is not returnable.
View return policy
Description
Plays a role in the formation of adherens junction and the polymerization of linear actin cables.Specifications
| FMN1 | |
| Polyclonal | |
| Unconjugated | |
| FMN1 | |
| FMN, FMN1, FMN1 Ens, formin 1, LD | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 342184 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| 0.29 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q68DA7 | |
| FMN1 | |
| FMN1 Fusion Protein Ag23059 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction