missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ FluxOR™ Red Potassium Ion Channel Assay

Product Code. p-7181122
Click to view available options
Quantity:
10 Plates
2 Plates
Unit Size:
Pack of 10
Pack of 2
This item is not returnable. View return policy

Product Code. 15632195

Brand: Invitrogen™ F20019

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

The FluxOR™ Red Potassium Ion Channel Assay utilizes a red fluorescent dye for the detection of potassium ion channel activity.

The assay takes advantage of the permeability of potassium channels to thallium ions. The red fluorescence (560/590 nm) of a thallium-sensitive indicator dye increases as the thallium concentration rises within the cell. The assay can be configured for either imaging or high throughput applications. Features include:

Large dynamic range—facilitates use in high throughput applications
Multiplexed enabled—red fluorescence does not interfere with GFP-expressing cells or other green fluorescent assays
Confirmation of results—green and red potassium ion channel assays can be used together for increased confidence in experimental results
Optimized format—96- or 384-well, suited for high throughput applications

Similar to the FluxOR™ II Green Potassium Ion Channel Assay, the FluxOR Red Potassium Ion Channel Assay makes use of the permeability of potassium channels to thallium. When potassium channels are opened by a stimulus, extracellular thallium flows down its concentration gradient into the cells, and channel or transporter activity is detected with a proprietary indicator dye pre-loaded into the cells that increases in cytosolic fluorescence in the presence of thallium. In this way, the fluorescence observed using the FluxOR Red assay is an indicator of any ion channel activity or transport process that allows thallium into cells. This fluorogenic signal quantitatively reflects the activity of ion channels and transporters that are permeant to thallium, including hERG, Kir2.1, and other pharmacologically important potassium channels.

The FluxOR Red Potassium Ion Channel Assay provides a concentrated thallium solution and all necessary buffers, allowing maximum target flexibility and ease of operation in a homogenous format that has been demonstrated with cells stably expressing hERG.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store contents at 2°C to 8°C and ≤-20°C.
For Use With (Application) Ion Channel Assay
Product Type Dye
Dye Type Potassium Indicator
Emission 560/590
Product Line FluxOR
Quantity 10 Plates
Shipping Condition Wet Ice
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.