Learn More
Fluxametamide, MedChemExpress
Description
Fluxametamide is an insecticide with wide spectrum, acts as an antagonist of GABA- and glutamate-gated chloride channels, with IC50 of 1.95 nM and 225 nM for M. domestica GABACls and GluCls.
Specifications
Specifications
| Chemical Name or Material | Fluxametamide |
| Physical Form | Solid |
| CAS | 928783-29-3 |
| Color | White |
| Quantity | 10 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C20H16Cl2F3N3O3 |
| Solubility Information | DMSO : 125 mg/mL (263.57 mM; Need ultrasonic) |
| SMILES | FC(F)(C1(C2=CC(Cl)=CC(Cl)=C2)CC(C3=CC(C)=C(C(N/C=N/OC)=O)C=C3)=NO1)F |
| Molecular Weight (g/mol) | 474.26 |
| Show More |