Learn More
Fluxametamide 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Product Code. 30317637
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30317637 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30317637 Supplier Dr. Ehrenstorfer Supplier No. DREA13874000AL100
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Fluxametamide
CAS 928783-29-3
Analyte or Component Names Fluxametamide
Shipping Condition +20°C
Molecular Formula C20 H16 Cl2 F3 N3 O3
Concentration 100 μg/mL
Certifications/Compliance ISO 17034
Synonym 4-[5-(3,5-Dichlorophenyl)-4,5-dihydro-5-(trifluoromethyl)-3-isoxazolyl]-N-[(methoxyamino)methylene]-2-methylbenzamide, 4-[5-(3,5-Dichlorophenyl)-5-trifluoromethyl-4,5-dihydroisoxazol-3-yl]-N-[(methoxyimino)methyl]-2-methylbenzamide, Fluxametamide
InChI Formula InChI=1S/C20H16Cl2F3N3O3/c1-11-5-12(3-4-16(11)18(29)26-10-27-30-2)17-9-19(31-28-17,20(23,24)25)13-6-14(21)8-15(22)7-13/h3-8,10H,9H2,1-2H3,(H,26,27,29)
SMILES CON=CNC(=O)c1ccc(cc1C)C2=NOC(C2)(c3cc(Cl)cc(Cl)c3)C(F)(F)F
IUPAC Name 4-[5-(3,5-dichlorophenyl)-5-(trifluoromethyl)-4H-1,2-oxazol-3-yl]-N-(methoxyiminomethyl)-2-methylbenzamide
Solvent or Matrix Acetonitrile
Molecular Weight (g/mol) 474.26
Quantity 1 mL
Formula Weight 473.0521 g/mol