Learn More
Fluvoxamine EP Impurity I, Mikromol™
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
Description
Fluvoxamine EP Impurity I
Specifications
Specifications
| Chemical Name or Material | (E)-N-[5-Methoxy-1-[4-(trifluoromethyl)phenyl]pentylidene]hydroxylamine |
| CAS | 61747-22-6 |
| Analyte or Component Names | (E)-N-[5-Methoxy-1-[4-(trifluoromethyl)phenyl]pentylidene]hydroxylamine |
| Shipping Condition | Room Temperature |
| Molecular Formula | C13 H16 F3 N O2 |
| Certifications/Compliance | ISO 9001 |
| Synonym | (E)-N-[5-Methoxy-1-[4-(trifluoro-methyl)phenyl]pentylidene]hydroxylamine, Fluvoxamine Maleate Imp. I (EP), 5-Methoxy-1-[4-(trifluoromethyl)phenyl]-1-pentanone Oxime |
| InChI Formula | InChI=1S/C13H16F3NO2/c1-19-9-3-2-4-12(17-18)10-5-7-11(8-6-10)13(14,15)16/h5-8,18H,2-4,9H2,1H3/b17-12+ |
| SMILES | COCCCC\C(=N/O)\c1ccc(cc1)C(F)(F)F |
| IUPAC Name | (NE)-N-[5-methoxy-1-[4-(trifluoromethyl)phenyl]pentylidene]hydroxylamine |
| Show More |