Learn More
Fluvastatin Ethyl Ester, Mikromol™
Description
Fluvastatin Ethyl Ester
Specifications
Specifications
| Chemical Name or Material | Fluvastatin Ethyl Ester |
| CAS | 615263-37-1 |
| Analyte or Component Names | Fluvastatin Ethyl Ester |
| Shipping Condition | Room Temperature |
| Molecular Formula | C26 H30 F N O4 |
| Certifications/Compliance | ISO 9001 |
| Synonym | (3R,5S,6E)-rel-7-[3-(4-Fluorophenyl)-1-(1-methylethyl)-1H-indol-2-yl]-3,5-dihydroxy-6-heptenoic acid ethyl ester |
| InChI Formula | InChI=1S/C26H30FNO4/c1-4-32-25(31)16-21(30)15-20(29)13-14-24-26(18-9-11-19(27)12-10-18)22-7-5-6-8-23(22)28(24)17(2)3/h5-14,17,20-21,29-30H,4,15-16H2,1-3H3/b14-13+/t20-,21-/m1/s1 |
| SMILES | CCOC(=O)C[C@H](O)C[C@H](O)\C=C\c1c(c2ccc(F)cc2)c3ccccc3n1C(C)C |
| IUPAC Name | ethyl (E,3R,5S)-7-[3-(4-fluorophenyl)-1-propan-2-ylindol-2-yl]-3,5-dihydroxyhept-6-enoate |
| Show More |