Learn More
Fluoxymesterone 1.0 mg/ml in Acetonitrile, LoGiCal
Specifications
Specifications
| Chemical Name or Material | Fluoxymesterone |
| CAS | 76-43-7 |
| Analyte or Component Names | Fluoxymesterone |
| Shipping Condition | Ice pack (-20°C) |
| Molecular Formula | C20 H29 F O3 |
| Certifications/Compliance | ISO 17034 |
| Synonym | Fluoxymesterone, 11β,17β-Dihydroxy-9α-fluoro-17α-methyl-4-androsten-3-one, 17α-Methyl-9α-fluoro-11β-hydroxytestosterone, 9-Fluoro-11β,17β-dihydroxy-17-methylandrost-4-en-3-one, 9α-Fluoro-11β,17β-dihydroxy-17-methyl-4-androsten-3-one, 9α-Fluoro-11β,17β-dihydroxy-17α-methyl-4-androsten-3-one, 9α-Fluoro-11β-hydroxy-17-methyltestosterone, 9α-Fluoro-11β-hydroxy-17α-methyltestosterone, Androfluorene, Androfluorone, Androsterolo, Fluotestin, Fluoximesterone, Flusteron, Flutestos, Halotestin, NSC 10704, NSC-12165, Neo-Ormonal, Ora Testryl, Oralsterone, Oratestin, Testoral, U 6040, Ultandren, Ultandrene, FLUOXYMESTERONE |
| InChI Formula | InChI=1S/C20H29FO3/c1-17-8-6-13(22)10-12(17)4-5-15-14-7-9-19(3,24)18(14,2)11-16(23)20(15,17)21/h10,14-16,23-24H,4-9,11H2,1-3H3/t14-,15-,16-,17-,18-,19-,20-/m0/s1 |
| Solution Type | Single Solution |
| SMILES | C[C@]1(O)CC[C@H]2[C@@H]3CCC4=CC(=O)CC[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]12C |
| Show More |