Learn More
Cytiva FluoroTrans™ W PVDF Transfer Membrane
Product Code. 17114721
Click to view available options
Dimensions (LxW):
10 x 15 cm
20 x 20 cm
26 cm x 3.3 m
7 x 9 cm
Unit Size:
1 roll
Pack of 10
Product Code. 17114721
Brand: Cytiva BSP0161
Only null left
Add to basket
Add to basket
FluoroTrans W membrane is optimized for Western transfer applications.
  • This membrane allows for sensitive protein detection with low background and very low protein burn-through
  • FluoroTrans W membrane is compatible with all total protein stains and immunodetection methods
TRUSTED_SUSTAINABILITY
Specifications
Packaging Type Roll
Description Non-sterile
Material FluoroTrans PVDF
Pore Size 0.2 μm
Dimensions (LxW) 26 cm x 3.3 m
Thickness 127 μm