Learn More
Fluorescein disodium salt
Product Code. 15464139
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15464139 100 g 100g
15474139 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15464139 Supplier Thermo Scientific Alfa Aesar Supplier No. J61549.22
  • ONLINE EXCLUSIVE PRICE
    580.00 NOK
    448.65 NOK / 100g
    Save 131.35 NOK
    22% Off
 Restricted Item
In Stock
Add to basket
Edge
Fluorescein disodium salt (uranin, fluorescite, acid yellow 73, CAS # 518-45-6) is the salt of the xanthene dye commonly used for fluorescent tracing. | CAS: 518-47-8 | C20H10Na2O5 | 376.28 g/mol
Fluorescein disodium salt is used as fluorescent tracer, has been utilized in studies as a drug model, as a penetrating dye . Water soluble fluorescent dye used in permeability and vascular perfusion studies due to its small molecular radius.1 Sodium fluorescein is used as a fluorescent tracer to study blood-brain barrier (BBB) and blood-spinal cord barrier (BSCB) permeability in rodent models. The dye was used as a probe substrate in the study of organic anion transporting polypeptide (OATP) mediated drug transport in hepatocytes.5 Sodium fluorescein was also used to monitor in situ solute transportation into bone through fluorescence recovery after photobleaching (FRAP). Additional reported uses include microscopy, as a gain medium in dye laser use and to detect latent blood stains in vitro. Common derivatives of Fluorescein include fluorescein isothiocyanate (FITC), eosin and Rose Bengal (sc-296278) and Rose Bengal sodium salt
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General Description
• Fluorescein disodium salt is a yellow-green phthalate indicator dye that is pH-sensitive
• It has a maximum absorbance of 494nm and maximum emission of 521nm
Application
• Fluorescein disodium salt can be used as a fluorescent protein labeler
• Fluorescein is also useful for the staining of cells and organelles
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 518-47-8
Molecular Formula C20H10Na2O5
Molecular Weight (g/mol) 376.28
MDL Number MFCD00167039
InChI Key NJDNXYGOVLYJHP-UHFFFAOYSA-L
Synonym Acid Yellow 73; C.I. 45350
IUPAC Name disodium 2-(6-oxido-3-oxo-3H-xanthen-9-yl)benzoate
SMILES [Na+].[Na+].[O-]C(=O)C1=CC=CC=C1C1=C2C=CC(=O)C=C2OC2=CC([O-])=CC=C12
Specifications
Color Brown-Red
Quantity 100 g
Beilstein 3833041
Merck Index 14,4159
Solubility Information Soluble in water at 1mg/ml
Formula Weight 376.28
Physical Form Powder
Chemical Name or Material Fluorescein disodium salt