Learn More
Cytiva Fluor Carbon Rubber O-ring
613.24 NOK - 771.00 NOK
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
10566540
|
Cytiva
19-7584-01 |
11.3 x 2.4mm |
771.00 NOK
Pack of 5 |
Estimated Shipment: 14-05-2024
Log in to see stock. |
|||||
12495600
|
Cytiva
28-9535-45 |
13.1 x 1.6mm |
613.24 NOK
Each |
Estimated Shipment: 14-05-2024
Log in to see stock. |
|||||
Specifications
O-Ring | |
Fluor Carbon Rubber |