Learn More
Flunixin-d3, TRC
Specifications
Specifications
| Chemical Name or Material | Flunixin D3 (methyl D3) |
| CAS | 1015856-60-6 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C14 2H3 H8 F3 N2 O2 |
| Quantity | 10 mg |
| Synonym | Flunixin D3 (methyl D3) |
| InChI Formula | InChI=1S/C14H11F3N2O2/c1-8-10(14(15,16)17)5-2-6-11(8)19-12-9(13(20)21)4-3-7-18-12/h2-7H,1H3,(H,18,19)(H,20,21)/i1D3 |
| SMILES | [2H]C([2H])([2H])c1c(Nc2ncccc2C(=O)O)cccc1C(F)(F)F |
| IUPAC Name | 2-[2-(trideuteriomethyl)-3-(trifluoromethyl)anilino]pyridine-3-carboxylic acid |
| Molecular Weight (g/mol) | 299.26 |
| Show More |