Learn More
Sampling Systems™ Fluid Point Sampler
Product Code. 15819371 Shop All Sampling Systems Products
Click to view available options
Capacity (Metric):
160 mL
300 mL
450 mL
Length (Metric):
600 mm
1000 mm
1500 mm
Unit Size:
Each
Product Code. 15819371
Brand: Sampling Systems™ 5551S1000
Only null left
Add to basket
Add to basket
Designed to allow rapid point sampling from drums, kegs and most containers. Sampling Systems™ Fluid Point Sampler is ideal for sampling pharmaceuticals, cosmetics and food stuffs.
  • Manufactured in 316L stainless steel with a silicone seal (FDA grade)
  • Available in a variety of lengths
  • Easy to clean
Specifications
Material Stainless Steel (316L)
Includes Silicone Seal
Capacity (Metric) 300 mL
Diameter (Metric) Outer 25 mm
Length (Metric) 1000 mm