Learn More
Thermo Scientific Chemicals Flucloxacillin, sodium salt, 95%, Thermo Scientific Chemicals
Shop All Thermo Scientific Chemicals ProductsDescription
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Type | Flucloxacillin Sodium Salt |
| Melting Point | 176.0°C to 178.0°C |
| Infrared Spectrum | Authentic |
| Loss on Drying | 4.5% max. |
| Specific Rotation | +158° to +168° (20°C, 589nm) (c=1, water) On dry substance |
| Quantity | 5 g |
| Flash Point | >110°C |
| Boiling Point | 677.0°C |
| IUPAC Name | sodium (2S,5R,6R)-6-[3-(2-chloro-6-fluorophenyl)-5-methyl-1,2-oxazole-4-amido]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate |
| Formula Weight | 475.86 |
| Show More |
Safety and Handling
Product Identifier
- Flucloxacillin, sodium salt
- Danger
- Respiratory sensitiser Category 1
- Skin sensitiser Category 1
- H317-May cause an allergic skin reaction.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .