Learn More
Invitrogen™ Attune™ Xenith™ 200-nm Primary Focus Fluid Filter
Description
The Attune Xenith 200-nm Primary Focus Fluid Filter is designed for use with the Attune Xenith Flow Cytometer. This filter prevents contaminants from entering the focusing fluid reservoir of the instrument. It is recommended that filters are replaced every 6&ndash:12 months, depending on usage.
Specifications
Specifications
| Quantity | 1 Ea. |
| For Use With (Equipment) | Attune Xenith Flow Cytometer |
| Product Type | Filter |