Learn More
Invitrogen™ Attune™ Xenith™ 200-nm Primary Focus Fluid Filter
Product Code. 30484828
Change view
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30484828 1 Ea. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30484828 Supplier Invitrogen™ Supplier No. 100114360S
This focusing fluid filter reduces background in your Attune Xenith Flow Cytometer by filtering contaminants over 200nm in size out of the focusing fluid as it enters the instrument.
The Attune Xenith 200-nm Primary Focus Fluid Filter is designed for use with the Attune Xenith Flow Cytometer. This filter prevents contaminants from entering the focusing fluid reservoir of the instrument. It is recommended that filters are replaced every 6&ndash:12 months, depending on usage.
TRUSTED_SUSTAINABILITY
Specifications
Quantity 1 Ea.
For Use With (Equipment) Attune Xenith Flow Cytometer
Product Type Filter
For Research Use Only. Not for use in diagnostic procedures.