Learn More
R&D Systems™ FlowTACS Flow Cytometry Apoptosis Detection Kit
Description
Includes
Streptavidin-Fluorescein and all the agents necessary for generating positive controls
Can be used for the detection of apoptosis using flow cytometry, direct visualization using a fluorescent microscope or for quantitation using a fluorometer
Specifications
Specifications
| Quantity | 60 Tests |
| Content And Storage | The product requires storage at both -20 to -70° C and 2 - 8° C. Consult the product insert for specific storage temperatures. Do not use past expiration date. |