Description
- The portable IKA FLOW is an attachment that forms a flow chemistry reactor in combination with your IKA magnetic stirrer RCT digital. In addition, it has been designed to be used with any fume hood gas source (with a 6 mm pressure tubing and an adaptor to typical fume hood gas supplies) such as compressed air or nitrogen.
- IKA FLOW's scope of delivery includes in addition to the attachment, a glass bottle, a mixing chip, support screws, three on-and-off valves, a heating block, four disc reactors, and a labeled tubing set.
- Pair IKA FLOW with the RCT digital stirrer hotplate.
- Scope of delivery
- IKA FLOW System
- Glass Bottle
- Heating Block
- CDR.PP standard (4 pcs.)
- Capillary Set
- Tubing set
Specifications
Specifications
| Volume (Metric) | 500 mL |
| Material | 304 Stainless Steel, Nylon (PA) |
| Weight (Metric) | 2.7 kg |
| For Use With (Equipment) | IKA Magnetic Stirrer RCT Digital |
| Includes | Glass Bottle, Mixing Chip, Support Screws, Three Valves, Heating Block, Four Disc Reactors, Labeled Tubing Set |