Learn More
Semadeni X5 Flow indicator SAN 90mm black
Brand:  Semadeni 1171
X5 Flow indicator SAN 90mm black
Product Code. 30277381
  • ONLINE EXCLUSIVE PRICE
    2427.30 NOK
    1155.91 NOK / Pack of 5
    Save 1271.39 NOK
    52% Off
Only null left
Estimated Shipment: 16-02-2026