Learn More
BOLA Flow Filters
Description
- Suitable for overpressure or vacuum, usable for example as added filter or as large-area in-line apparatus in a line system.
- Suitable for temperatures up to +160°C.
- The optionally available filtering membranes can be exchanged easily.
- Tubing can be connected to GL threads by means of the included laboratory screw joints.
- The filters are produced without plasticisers and have an almost universal chemical resistance.
- They do not release any trace elements into the filtrate.
- Due to the non-adhesive surface, they are easy to clean and can be reused.
Specifications
Specifications
| Autoclavable | Autoclavable |
| Disposable | Reusable |
| Sample Volume | 52 qcm |
| Sterility | Non-sterile |
| Open Area | 52 qcm |
| Diameter (Metric) | 8 mm |