Learn More
BOLA Flow Filters
Product Code. 10767332
Change view
Click to view available options
:
8 mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10767332 8 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10767332 Supplier BOLA Supplier No. N167024
Only null left
Add to basket
Add to basket
Flow Filters Filter made of PTFE, laboratory screw joints made of PPS for tubing
  • Suitable for overpressure or vacuum, usable for example as added filter or as large-area in-line apparatus in a line system.
  • Suitable for temperatures up to +160°C.
  • The optionally available filtering membranes can be exchanged easily.
  • Tubing can be connected to GL threads by means of the included laboratory screw joints.
  • The filters are produced without plasticisers and have an almost universal chemical resistance.
  • They do not release any trace elements into the filtrate.
  • Due to the non-adhesive surface, they are easy to clean and can be reused.
Specifications
Autoclavable Autoclavable
Disposable Reusable
Sample Volume 52 qcm
Sterility Non-sterile
Open Area 52 qcm
Diameter (Metric) 8 mm