Learn More
OHAUS™ Flow Control Clamp
Product Code. 15752269 Shop All Ohaus Products
Click to view available options
Material:
Nickel-plated
Stainless Steel
Unit Size:
Each
Product Code. 15752269
Brand: OHAUS™ 30392221
Only null left
Add to basket
Add to basket
OHAUS™ Flow Control Clamp in a variety of styles, materials, and sizes.
  • Designed to securely grip and position laboratory apparatus
  • Extension arms with a 360° rotation allow placement of apparatus at various distances from lab-frame without compromising experiment integrity
  • Swivel and fixed position clamps hold apparatus near the lab-frame or stand, and have a built-in integral holder for attaching to lab-frames or apparatus
  • Available as single or dual-adjust in zinc or stainless steel
TRUSTED_SUSTAINABILITY
Specifications
Type Multi-purpose Clamp
Material Stainless Steel
Description Clamp, Flow Control, Hosecock, CLF-HKRGS
Height (Metric) 62 mm
Height (English) 2.44 in.
Length (Metric) 37 mm
Size (Metric) Grip 0 to 17 mm
Width (English) 1.46 in.
Width (Metric) 37 mm
Jaw Type Hosecock