Learn More
OHAUS™ Flow Control Clamp
Shop All Ohaus Products
Click to view available options
Material:
Nickel-plated
Stainless Steel
Unit Size:
Each
Description
- Designed to securely grip and position laboratory apparatus
- Extension arms with a 360° rotation allow placement of apparatus at various distances from lab-frame without compromising experiment integrity
- Swivel and fixed position clamps hold apparatus near the lab-frame or stand, and have a built-in integral holder for attaching to lab-frames or apparatus
- Available as single or dual-adjust in zinc or stainless steel
Specifications
Specifications
| Type | Multi-purpose Clamp |
| Material | Nickel-plated |
| Description | Clamp, Flow Control, Hosecock, CLF-HKRGZ |
| Height (Metric) | 62 mm |
| Height (English) | 2.44 in. |
| Length (Metric) | 37 mm |
| Size (Metric) Grip | 0 to 17 mm |
| Width (English) | 1.46 in. |
| Width (Metric) | 37 mm |
| Jaw Type | Hosecock |