Learn More
Macherey-Nagel™ Florosil™ -Standard
Shop All Macherey-Nagel Products
Click to view available options
Quantity:
1 kg
5 kg
Unit Size:
1kg
5kg
Specifications
Specifications
| Particle Size | 0.15 to 0.25 mm |
| Quantity | 5 kg |
| Type | Florosil-Standard |
| Material | Magnesia Silica |