Learn More
Tork Floor Stand Dispenser W1
Product Code. 12827880 Shop All Tork Products
Change view
Click to view available options
Color:
Red, Smoke
Translucent Blue
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
12827880 Translucent Blue Each
30243717 Red, Smoke Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 12827880 Supplier Tork Supplier No. 652000
  • ONLINE EXCLUSIVE PRICE
    2330.00 NOK
    2296.77 NOK / Pack of 1
    Save 33.23 NOK
    1% Off
View all promotions
Estimated Shipment: 25-11-2026
The Tork floor stand dispenser in Performance design is developed for safety and stability with its reliable tear-off and easy-load features.
  • Ideal for a varied workflow - you can place our mobile dispenser where you need it.
  • Cut back on maintenance with our high-capacity stand designed for easy filling and refilling.
  • Save precious time by adding a matching bin liner-holder to the stand.
  • Compatible with a variety of Tork wiping papers and cleaning cloths, so you can pick the right product for your workspace.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Floor Stand Wiper Dispenser
Color Translucent Blue
Alternate Part Number 7322540430561
Length (Metric) 64.6 cm
Width (Metric) 53 cm
Height (Metric) 100.6 cm
Holds Wipes
Material Metal, Plastic
Mounting Freestanding
No. of Compartments 1
Product Style Front-loading with Opening
Model Performance