Learn More
Scotch-Brite™ Floor Pads, White
Click to view available options
Diameter (Metric):
280 mm
305 mm
330 mm
355 mm
380 mm
406 mm
432 mm
460 mm
480 mm
505 mm
530 mm
Unit Size:
Pack of 5
Beschreibung
- Used for light cleaning to help remove soil and scuff marks with minimal dulling of finish
- Highly conformable fibers provide maximum pad-to-floor contact, delivering outstanding cleaning results
- Reversible and washable polishing pad resists clogging and loading
- Uniform distribution of mineral abrasives throughout whole pad and not just on the surface
- Delivers consistent performance with long floor pad life
Spezifikation
Spezifikation
| Product Type | Floor Pads |
| Color | White |
| Diameter (Metric) | 280 mm |
| For Use With (Application) | Dry Polishing and Buffing Unprotected Floors |
| For Use With (Equipment) | Standard Speed Rotary Floor Machines |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur