Learn More
Scotch-Brite™ Floor Pads, White
Product Code. 17188800
Change view
Click to view available options
Diameter (Metric):
280 mm
305 mm
330 mm
355 mm
380 mm
406 mm
432 mm
460 mm
480 mm
505 mm
530 mm
Unit Size:
Pack of 5
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) unitSize
17188800 280 mm Pack of 5
17168810 305 mm Pack of 5
17148810 530 mm Pack of 5
17178810 330 mm Pack of 5
17178820 355 mm Pack of 5
17188820 380 mm Pack of 5
17108820 406 mm Pack of 5
17168820 432 mm Pack of 5
17118820 460 mm Pack of 5
17158820 480 mm Pack of 5
17128820 505 mm Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
Product Code. 17188800 Supplier Scotch-Brite™ Supplier No. 7100159878
  • ONLINE EXCLUSIVE PRICE
    802.00 NOK
    503.38 NOK / Pack of 5
    Save 298.62 NOK
    37% Off
Estimated Shipment: 29-09-2026
Add to basket
Add to basket
Ideal for dry polishing and buffing unprotected floors with standard speed rotary floor machines. These pads are also suitable for light spray cleaning. They enhance floor shine without scratching or damaging protected floors.
  • Used for light cleaning to help remove soil and scuff marks with minimal dulling of finish
  • Highly conformable fibers provide maximum pad-to-floor contact, delivering outstanding cleaning results
  • Reversible and washable polishing pad resists clogging and loading
  • Uniform distribution of mineral abrasives throughout whole pad and not just on the surface
  • Delivers consistent performance with long floor pad life
TRUSTED_SUSTAINABILITY
Specifications
Product Type Floor Pads
Color White
Diameter (Metric) 280 mm
For Use With (Application) Dry Polishing and Buffing Unprotected Floors
For Use With (Equipment) Standard Speed Rotary Floor Machines