Learn More
Scotch-Brite™ Floor Pads, Green
Product Code. 17108810
Change view
Click to view available options
Diameter (Metric):
254 mm
280 mm
305 mm
330 mm
355 mm
380 mm
406 mm
432 mm
460 mm
480 mm
505 mm
530 mm
Unit Size:
Pack of 25
Pack of 5
Product Code. Diameter (Metric) unitSize
17108810 406 mm Pack of 5
17168800 254 mm Pack of 5
17198810 280 mm Pack of 5
17148820 505 mm Pack of 5
17138810 530 mm Pack of 5
17178800 305 mm Pack of 5
17138820 330 mm Pack of 5
17158810 355 mm Pack of 5
17198800 380 mm Pack of 5
17118810 432 mm Pack of 5
17128810 460 mm Pack of 25
17277206 460 mm Pack of 5
17188810 480 mm Pack of 5
13 options
Product Code. 17108810
Brand: Scotch-Brite™ 7100159880
Only null left
Add to basket
Add to basket
For the efficient, intermediate cleaning of heavily soiled floors with or without a coating. For use with single disc machines or automatic floor cleaning machines with slow speed.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Floor Pad
Color Green
Diameter (Metric) 406 mm
For Use With (Application) Scrubbing
For Use With (Equipment) Single Disc Machines or Slow Speed Automatic Floor Cleaning Machines
Style Round