Learn More
Scotch-Brite™ Floor Pads, Green
Product Code. 17168800
Change view
Click to view available options
Diameter (Metric):
254 mm
280 mm
305 mm
330 mm
355 mm
380 mm
406 mm
432 mm
460 mm
480 mm
505 mm
530 mm
Unit Size:
Pack of 25
Pack of 5
13 product options available for selection
Product selection table with 13 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) unitSize
17168800 254 mm Pack of 5
17198810 280 mm Pack of 5
17148820 505 mm Pack of 5
17138810 530 mm Pack of 5
17178800 305 mm Pack of 5
17138820 330 mm Pack of 5
17158810 355 mm Pack of 5
17198800 380 mm Pack of 5
17108810 406 mm Pack of 5
17118810 432 mm Pack of 5
17128810 460 mm Pack of 25
17277206 460 mm Pack of 5
17188810 480 mm Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 13 options available.
13 options
Product Code. 17168800 Supplier Scotch-Brite™ Supplier No. 7100159535
  • ONLINE EXCLUSIVE PRICE
    657.00 NOK
    409.53 NOK / Pack of 5
    Save 247.47 NOK
    37% Off
Estimated Shipment: 29-09-2026
Add to basket
Add to basket
For the efficient, intermediate cleaning of heavily soiled floors with or without a coating. For use with single disc machines or automatic floor cleaning machines with slow speed.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Floor Pad
Color Green
Diameter (Metric) 254 mm
For Use With (Application) Scrubbing
For Use With (Equipment) Single Disc Machines or Slow Speed Automatic Floor Cleaning Machines
Style Round