Description
Optimize readings and temperature accuracy for a variety of applications with these probes. Cole-Parmer™ Digi-Sense™ Flexible Insulated-Wire Thermocouple Probes are ideal for high-temperature measurements and can be used with liquids, gases and semisolids.
All insulated-wire probes include straight insulated cable with no handle. Fiberglass and ceramic insulated probes have exposed junctions; use for high-temperature applications. Exposed junction probes are not recommended for use with aqueous solutions (type J exposed-junction probes will rust when exposed to moisture).
- Each probe comes with color coded ANSI mini-connectors: type J-black, type K-yellow, type T-blue, and type E-purple.
- Ceramic Fiber-Insulated Wire Probes for high-temperature measurements.
- General-Purpose Probes are designed for use with liquids, gases and semisolids.
- ABS handles provide a light weight design for ease of use along with durability
- Probe handles feature a strain relief spring that protects the cable connection against damage due to repeated flexing and tugging
Specifications
Specifications
| Certifications/Compliance | ANSI |
| For Use With (Equipment) | Temperature Meters |
| Temperature (Metric) | -150°C to +1100°C |
| Type | Thermocouple Probe |
| For Use With (Application) | High Temperature Environments or Pressurized Chambers |
| Material | Stainless-steel Handle |
| Style | Type-K Pipe-Ftg Probe Mini Conn |
| Length (Metric) | 10.16 cm |