Learn More
OHAUS™ Flex System Clamp
Product Code. 15762429 Shop All Ohaus Products
Click to view available options
Length (Metric) Arm:
305 mm
457 mm
Includes:
Base Plate
Lab Frame Clamp
Shelf Clamp
Unit Size:
Each
Product Code. 15762429
Brand: OHAUS™ 30400014
Only null left
Add to basket
Add to basket
OHAUS™ Flex System Clamp is designed to securely grip and position a variety of laboratory glassware.
  • Corrosion-resistant stainless steel clamps provide excellent chemical resistance, durability, and can be autoclaved. Nickel-plated zinc clamps offer high-tensile strength at an economical price.
  • Prevent accidents in the lab! Versatile multi-purpose LabJaws clamps securely hold glassware and irregularly-shaped objects. Select from a variety of grip sizes, prong styles and adjustment types.
  • Choose from a variety of specialized LabJaws clamps with adjustable features to provide an exact fit for lab ware such as thermometers, thermocouples, columns, funnels, and round bottom flasks.
TRUSTED_SUSTAINABILITY
Specifications
Type Flex System Clamp
Material Nickel-plated
Includes Base Plate
Description Clamp, Specialty, Flex, CLS-FLEXSL
Diameter (Metric) 13 mm
Height (Metric) 13 mm
Height (English) 0.5 in.
Length (English) 5 in.
Length (English) Arm 18 in.
Length (Metric) 127 mm
Length (Metric) Arm 457 mm
Width (English) 5 in.
Width (Metric) 127 mm
Jaw Type 3-Prong Dual Adjust