Learn More
Grant Instruments™ Multi-level Flat Platform Tower With Non-Slip Mat for PSU20i
Product Code. 12914035
Click to view available options
Description:
2-level
3-level
4-level
Unit Size:
Each
Product Code. 12914035
Brand: Grant Instruments™ PP202
Only null left
Add to basket
Add to basket
Flat shaker platform with multiple levels for use with the Grant PSU-20i orbital shaker. Black rubber pads on each platform provide a nonslip surface for vessels on the platform
  • For use with PSU 20i Orbital Shaking Platform
  • 2, 3, or 4 levels
  • Black rubber pads provide nonslip surface for vessels on the platform.
TRUSTED_SUSTAINABILITY
Specifications
Material Rubber
Dimensions (L x W) 480 x 380 mm
Description 2-level
For Use With (Equipment) PSU-20i
No. of Shelves 2-level