Learn More
Thermo Scientific™ Flat PCR Caps, strips of 12
Product Code. p-7184505
Click to view available options
Color:
Natural
Product Type:
Flat Cap, 12-Cap Strip
PCR Plate Cap Strip
Unit Size:
Pack of 200
Pack of 80
Product Code. 12109840
Brand: Thermo Scientific™ AB0851
Only null left
Add to basket
Add to basket
Domed cap strips for PCR applications for use with Thermo Scientific 96-well PCR plates and strip tubes.
Ensure proper sealing on all PCR strip tubes and 96-well PCR plates with precision molded Thermo Scientific™ PCR Plate Cap Strips.
Flat PCR Cap Strips are compatible with 96-well PCR plates and standard and low profile strip tubes. The 12 strips have large end tabs for easy strip removal.
Highlights:
  • Suitable for use with all 96-well PCR plates as well as standard and low profile strip tubes
  • Large end tabs for strip removal (12 strips only)
Certifications:
Certified free from human DNA, RNases, and DNases.
TRUSTED_SUSTAINABILITY
Specifications
Color Natural
Certifications/Compliance Certified DNA/RNase Free
Material Polypropylene
No. of Wells 12
Quantity 25 packs of 8
For Use With (Application) Thermal Cyclers
Format 200 Strip Caps
For Use With (Equipment) Thermal Cyclers
Product Type PCR Plate Cap Strip
Certified RNase/DNase-free Yes
Plate Compatibility 96-well Plate