Learn More
Heidolph™ Flat Hood
Product Code. 10674131 Shop All Heidolph Instruments Products
Change view
Click to view available options
For Use With (Equipment):
Incubating Shakers
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
10674131 Incubating Shakers Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10674131 Supplier Heidolph™ Supplier No. 5499004000
Estimated Shipment: 29-12-2026
For use as an add on to incubating shakers
With a height of 163 mm, the flat incubation hood is perfectly suited for multi-well plates, culture flasks and Erlenmeyer flasks with a volume up to 100 mL.
TRUSTED_SUSTAINABILITY
Specifications
Holds Multi-well plates, culture flasks and Erlenmeyer flasks with a volume up to 100 mL
Product Type Incubation Hood
Height (Metric) 163 mm
For Use With (Equipment) Incubating Shakers