Description
- Flat flange allows the lid to be securely attached to the reactor body and ensures a reliable seal to prevent leakage
- Compatible according to DIN 12214 (laboratory flat flange) and compatible with glass reactors for seamless integration
- Effortless and hassle-free to assemble, saving time and effort
- Flexibly expand our products to fit your specific needs and meet future needs
- Made entirely of PTFE, for universal chemical resistance to a wide range of harsh chemicals, increasing application versatility and reliability
- Equipped with practical attachments that allow easy connection and use of existing equipment that has ground or GL threads
- Important component for the safe and reliable connection of pipes and vessels in various industrial applications to prevent leaks and keep equipment running smoothly
Specifications
Specifications
| Product Type | Flat Flange |
| For Use With (Application) | Connection of Pipes and Vessels, Preventing Leaks |
| For Use With (Equipment) | Glass Reactors |
| Includes | 1 x NPT 1/2 in. (f) Centered, 3 x NPT 1/2 in. (f) Beside Center |
| Joint Type | DN 60 |
| Material | PTFE |