Learn More
Lenz Laborglasinstrumente™ Flat Flange Lid with Center Neck and Two Angled Side Necks
Product Code. 11582354
Change view
Click to view available options
Height (Metric):
75 mm
85 mm
95 mm
110 mm
125 mm
150 mm
165 mm
Includes:
Laboratory Flange 100
Laboratory Flange 140
Laboratory Flange 150
Laboratory Flange 200
Laboratory Flange 60
Unit Size:
Each
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Height (Metric) Includes Unit Size
11582354 165 mm Laboratory Flange 200 Each
10311560 110 mm Laboratory Flange 100 Each
10775695 95 mm Laboratory Flange 100 Each
11562354 150 mm Laboratory Flange 140 Each
11552354 125 mm Laboratory Flange 150 Each
11572354 150 mm Laboratory Flange 200 Each
11542354 75 mm Laboratory Flange 60 Each
10192454 85 mm Laboratory Flange 60 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Product Code. 11582354 Supplier Lenz Laborglasinstrumente™ Supplier No. 06103564
  • ONLINE EXCLUSIVE PRICE
    7940.00 NOK
    6646.73 NOK / Pack of 1
    Save 1293.27 NOK
    16% Off
Estimated Shipment: 02-02-2027
Add to basket
Add to basket
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Spezifikation
Material Borosilicate Glass, Duran Glass
For Use With (Application) Reaction vessels
Height (Metric) 165 mm
Includes Laboratory Flange 200
Product Type Flange lid