Learn More
Lenz Laborglasinstrumente™ Flat Flange Lid with Center Neck, Two Angled Side Necks, and One Vertical Side Neck
Click to view available options
Closure Size:
Center Neck (NS): 14/23
Center Neck (NS): 29/32
Center Neck (NS): 45/40
Includes:
Laboratory Flange 100
Laboratory Flange 150
Laboratory Flange 200
Laboratory Flange 60
Unit Size:
Each
Specifications
Specifications
| Material | Borosilicate Glass, Duran Glass |
| For Use With (Application) | Reaction vessels |
| Includes | Laboratory Flange 150 |
| Product Type | Flange lid |
| Closure Size | Center Neck (NS): 45/40 |