Learn More
Lenz Laborglasinstrumente™ Flat Flange Lid with Center Neck, Three Angled Side Necks, and One Vertical Side Neck
2809.00 NOK - 8368.70 NOK
Specifications
Material | Borosilicate Glass, Duran Glass |
---|---|
For Use With (Application) | Reaction vessels |
Product Type | Flange lid |
Product Code | Brand | Includes | Closure Size | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Includes | Closure Size | Price | Quantity & Availability | ||||
11322503
|
Lenz Laborglasinstrumente™
06105241 |
Laboratory Flange 100 | Center Neck (NS): 29/32 |
2809.00 NOK
Each |
Estimated Shipment: 05-07-2024
Log in to see stock. |
||||
10536084
|
Lenz Laborglasinstrumente™
06105264 |
Laboratory Flange 100 | Center Neck (NS): 45/40 |
3439.70 NOK
Each |
Estimated Shipment: 05-07-2024
Log in to see stock. |
||||
10793062
|
Lenz Laborglasinstrumente™
06105544 |
Laboratory Flange 200 | Center Neck (NS): 29/32 |
8268.00 NOK
Each |
Estimated Shipment: 05-07-2024
Log in to see stock. |
||||
11562364
|
Lenz Laborglasinstrumente™
06105464 |
Laboratory Flange 150 | Center Neck (NS): 45/40 |
4796.50 NOK
Each |
Estimated Shipment: 05-07-2024
Log in to see stock. |
||||
11572364
|
Lenz Laborglasinstrumente™
06105564 |
Laboratory Flange 200 | Center Neck (NS): 45/40 |
8368.70 NOK
Each |
Estimated Shipment: 05-07-2024
Log in to see stock. |
||||
Specifications
Borosilicate Glass, Duran Glass | |
Reaction vessels | |
Flange lid |