Learn More
SCAT Flat Canisters
Product Code. p-9356000 Shop All SCAT Europe Products
Click to view available options
Capacity (Metric):
8 L
20 L
Closure Size:
63B
S70/71
Dimensions (L x W x H):
283 x 376 x 292 mm
375 x 305 x 165 mm
Unit Size:
Each
Product Code. 10561001
Brand: SCAT 107709
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Due to their flat design, SCAT flat canisters have been specially developed for use in confined spaces such as cupboards, shelves, drawers or refrigerators, as they are structurally low in height.
  • Made of either robust HDPE or PP and therefore resistant to all solvents commonly used in laboratories
  • Available with B63 or S70/71 threads and can be adapted to many other thread sizes by means of an adapter
  • Available either as compact 8 L canister or as more voluminous 20 L canister
SCAT flat canisters are an ideal way of not having to dispense with safe storage even when space is limited, as they can be integrated into laboratory furniture to save space. SCAT flat pack canisters are also optimally available with UN-Y approval for the storage of hazardous goods or with an integrated drain cock for safe emptying.
Specifications
Autoclavable Not Autoclavable
Capacity (English) 2 gal.
Capacity (Metric) 8 L
Closure Size 63B
Color White (Translucent)
Description Flat can with drain valve, 8 L, B63, PP, dimensions: (W x H x D): 375 x 165 x 305 mm, autoclavable
Dimensions (L x W x H) 375 x 305 x 165 mm
Includes with drain valve, autoclavable
Material Polypropylene
Mouth Style B63 Thread
Product Type Canister